Assay Details
| Gene Symbol: Atac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144394.2 | NP_001137866.1 | 6 - 7 | 1330 | 68 | |
| NM_001259602.2 | NP_001246531.1 | 8 - 9 | 1453 | 68 | ||
| NM_001274275.1 | NP_001261204.1 | 7 - 8 | 1351 | 68 | ||
| NM_167828.4 | NP_728520.2 | 7 - 8 | 1483 | 68 | ||
More Information
Set Membership: |
|

Atac3
Genomic Map