Assay Details
| Gene Symbol: Atac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144394.2 | NP_001137866.1 | 7 - 8 | 1405 | 77 | |
| NM_001259602.2 | NP_001246531.1 | 9 - 10 | 1528 | 77 | ||
| NM_001274275.1 | NP_001261204.1 | 8 - 9 | 1426 | 77 | ||
| NM_167828.4 | NP_728520.2 | 8 - 9 | 1558 | 77 | ||
More Information
Set Membership: |
|

Atac3
Genomic Map