Assay Details
| Gene Symbol: fs(1)N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130545.3 | NP_569901.1 | 2 - 3 | 2241 | 78 | |
| GenBank mRNA | AY118366.1 | - | 2 - 3 | 2229 | 78 | |
More Information
Set Membership: |
|

fs(1)N
Genomic Map