Assay Details
| Gene Symbol: fs(1)N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130545.3 | NP_569901.1 | 3 - 4 | 2364 | 56 | |
| GenBank mRNA | AY118366.1 | - | 3 - 4 | 2352 | 56 | |
More Information
Set Membership: |
|

fs(1)N
Genomic Map