Assay Details
| Gene Symbol: fs(1)N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130545.3 | NP_569901.1 | 6 - 7 | 4485 | 70 | |
| GenBank mRNA | AY118366.1 | - | 6 - 7 | 4473 | 70 | |
More Information
Set Membership: |
|

fs(1)N
Genomic Map