Assay Details
| Gene Symbol: mei-38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347750.1 | NP_001334676.1 | 1 - 2 | 229 | 64 | |
| NM_130566.3 | NP_569922.1 | 1 - 2 | 229 | 64 | ||
| GenBank mRNA | BT044309.1 | - | 1 - 2 | 198 | 64 | |
More Information
Set Membership: |
|

mei-38
Genomic Map