Assay Details
| Gene Symbol: Hr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272227.1 | NP_001259156.1 | 7 | 1658 | 100 | |
| NM_001272229.1 | NP_001259158.1 | 1 | 1649 | 100 | ||
| NM_001272233.2 | NP_001259162.1 | 6 | 2275 | 100 | ||
| NM_001272234.1 | NP_001259163.1 | 7 | 2329 | 100 | ||
More Information
Set Membership: |
|

Hr4
Genomic Map