Assay Details
| Gene Symbol: Hr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038734.2 | NP_001033823.1 | 10 - 11 | 5407 | 81 | |
| NM_001272227.1 | NP_001259156.1 | 11 - 12 | 6390 | 81 | ||
| NM_001272228.1 | NP_001259157.1 | 6 - 7 | 5421 | 81 | ||
| NM_001272229.1 | NP_001259158.1 | 5 - 6 | 6378 | 81 | ||
| NM_001272230.1 | NP_001259159.1 | 11 - 12 | 5600 | 81 | ||
| NM_001272231.1 | NP_001259160.1 | 10 - 11 | 5410 | 81 | ||
| NM_001272232.1 | NP_001259161.1 | 10 - 11 | 5410 | 81 | ||
| NM_001272233.2 | NP_001259162.1 | 10 - 11 | 7007 | 81 | ||
| NM_001272234.1 | NP_001259163.1 | 11 - 12 | 7061 | 81 | ||
| GenBank mRNA | AY971884.1 | - | 4 - 5 | 4483 | 81 | |
More Information
Set Membership: |
|

Hr4
Genomic Map