Assay Details
| Gene Symbol: Pi4KIIIalpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272262.2 | NP_001259191.1 | 5 - 6 | 2147 | 70 | |
| NM_130658.3 | NP_570014.3 | 4 - 5 | 2087 | 70 | ||
More Information
Set Membership: |
|

Pi4KIIIalpha
Genomic Map