Assay Details
| Gene Symbol: Ugalt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144677.3 | NP_001138149.1 | 1 - 2 | 462 | 70 | |
| NM_001297904.1 | NP_001284833.1 | 2 - 3 | 472 | 70 | ||
| NM_130663.4 | NP_570019.1 | 2 - 3 | 365 | 70 | ||
| GenBank mRNA | AB055493.1 | - | 2 - 3 | 357 | 70 | |
| AF397530.1 | - | 2 - 3 | 345 | 70 | ||
| BT003169.1 | - | 2 - 3 | 396 | 70 | ||
More Information
Set Membership: |
|

Ugalt
Genomic Map