Assay Details
| Gene Symbol: eIF2Bbeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130664.4 | NP_570020.1 | 4 - 5 | 1132 | 67 | |
| GenBank mRNA | AF289101.1 | - | 4 - 5 | 985 | 67 | |
| AJ344147.1 | - | 4 - 5 | 1080 | 67 | ||
| BT023929.1 | - | 4 - 5 | 1127 | 67 | ||
More Information
Set Membership: |
|

eIF2Bbeta
Genomic Map