Assay Details
| Gene Symbol: ttm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316628.1 | NP_001303557.1 | 4 - 5 | 1238 | 92 | |
| NM_130671.3 | NP_570027.1 | 4 - 5 | 1238 | 92 | ||
| GenBank mRNA | AY070535.1 | - | 4 - 5 | 1214 | 92 | |
More Information
Set Membership: |
|

ttm50
Genomic Map