Assay Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297911.1 | NP_001284840.1 | 3 - 4 | 505 | 61 | |
| NM_001297912.1 | NP_001284841.1 | 4 - 5 | 824 | 61 | ||
| NM_130709.2 | NP_570065.2 | 4 - 5 | 824 | 61 | ||
| GenBank mRNA | AF468957.1 | - | 4 - 5 | 586 | 61 | |
| AY118453.1 | - | 4 - 5 | 824 | 61 | ||
More Information
Set Membership: |
|

Gas8
Genomic Map