Assay Details
| Gene Symbol: VhaAC39-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130724.4 | NP_570080.1 | 2 - 3 | 884 | 74 | |
| GenBank mRNA | AY069530.1 | - | 2 - 3 | 873 | 74 | |
More Information
Set Membership: |
|

VhaAC39-1
Genomic Map