Assay Details
| Gene Symbol: CG42541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038740.3 | NP_001033829.2 | 3 - 4 | 1465 | 121 | |
| NM_001272296.1 | NP_001259225.1 | 3 - 4 | 1465 | 121 | ||
| NM_206622.4 | NP_996345.3 | 3 - 4 | 1465 | 121 | ||
More Information
Set Membership: |
|

CG42541
Genomic Map