Assay Details
| Gene Symbol: MED16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137806.4 | NP_611650.2 | 1 | 2655 | 111 | |
| GenBank mRNA | BT025233.1 | - | 1 | 2647 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED16| Gene Symbol: MED16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137806.4 | NP_611650.2 | 1 | 2655 | 111 | |
| GenBank mRNA | BT025233.1 | - | 1 | 2647 | 111 | |
Set Membership: |
|