Assay Details
| Gene Symbol: Oatp58Da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137813.5 | NP_611657.2 | 3 - 4 | 769 | 66 | |
| GenBank mRNA | BT030425.1 | - | 3 - 4 | 761 | 66 | |
More Information
Set Membership: |
|

Oatp58Da
Genomic Map