Assay Details
| Gene Symbol: Oatp58Da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137813.5 | NP_611657.2 | 4 - 5 | 1610 | 74 | |
| GenBank mRNA | BT030425.1 | - | 4 - 5 | 1602 | 74 | |
More Information
Set Membership: |
|

Oatp58Da
Genomic Map