Assay Details
| Gene Symbol: Ugt317A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299805.1 | NP_001286734.1 | 3 - 4 | 368 | 78 | |
| NM_137850.4 | NP_611694.2 | 3 - 4 | 368 | 78 | ||
| GenBank mRNA | AY128460.1 | - | 3 - 4 | 367 | 78 | |
More Information
Set Membership: |
|

Ugt317A1
Genomic Map