Assay Details
| Gene Symbol: Klp59D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137918.3 | NP_611762.1 | 1 | 2452 | 136 | |
| GenBank mRNA | BT150331.1 | - | 1 | 2452 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klp59D| Gene Symbol: Klp59D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137918.3 | NP_611762.1 | 1 | 2452 | 136 | |
| GenBank mRNA | BT150331.1 | - | 1 | 2452 | 136 | |
Set Membership: |
|