Assay Details
| Gene Symbol: wmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103966.2 | NP_001097436.1 | 2 - 3 | 558 | 100 | |
| NM_137960.3 | NP_611804.1 | 1 - 2 | 503 | 100 | ||
| GenBank mRNA | AY061000.1 | - | 1 - 2 | 469 | 100 | |
More Information
Set Membership: |
|

wmd
Genomic Map