Assay Details
| Gene Symbol: ItgaPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137964.2 | NP_611808.2 | 6 - 7 | 3002 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ItgaPS5| Gene Symbol: ItgaPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137964.2 | NP_611808.2 | 6 - 7 | 3002 | 74 | |
Set Membership: |
|