Assay Details
| Gene Symbol: Nxt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137989.3 | NP_611833.1 | 2 - 3 | 122 | 81 | |
| GenBank mRNA | AF156959.1 | - | 1 - 2 | 95 | 81 | |
| AY071250.1 | - | 2 - 3 | 121 | 81 | ||
| KX531837.1 | - | 1 - 2 | 95 | 81 | ||
More Information
Set Membership: |
|

Nxt1
Genomic Map