Assay Details
| Gene Symbol: DNAlig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137999.3 | NP_611843.2 | 5 - 6 | 1720 | 69 | |
| GenBank mRNA | BT016103.1 | - | 5 - 6 | 1695 | 69 | |
More Information
Set Membership: |
|

DNAlig1
Genomic Map