Assay Details
| Gene Symbol: thoc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138012.3 | NP_611856.1 | 5 - 6 | 1687 | 62 | |
| GenBank mRNA | AJ620304.1 | - | 5 - 6 | 1614 | 62 | |
| AY061305.1 | - | 5 - 6 | 1639 | 62 | ||
More Information
Set Membership: |
|

thoc5
Genomic Map