Assay Details
| Gene Symbol: cN-IIIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138051.4 | NP_611895.1 | 1 | 1388 | 84 | |
| GenBank mRNA | AY061358.1 | - | 1 | 1344 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cN-IIIB| Gene Symbol: cN-IIIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138051.4 | NP_611895.1 | 1 | 1388 | 84 | |
| GenBank mRNA | AY061358.1 | - | 1 | 1344 | 84 | |
Set Membership: |
|