Assay Details
| Gene Symbol: CG13579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274240.2 | NP_001261169.1 | 5 - 6 | 1102 | 67 | |
| NM_138073.4 | NP_611917.2 | 5 - 6 | 1102 | 67 | ||
| Gene Symbol: sta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT029094.1 | - | 1086 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT029094.1 transcripts.
Set Membership: |
|

CG13579
Genomic Map