Assay Details
| Gene Symbol: CG13579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274240.2 | NP_001261169.1 | 8 - 9 | 1811 | 59 | |
| NM_138073.4 | NP_611917.2 | 8 - 9 | 1811 | 59 | ||
More Information
Set Membership: |
|

CG13579
Genomic Map