Assay Details
| Gene Symbol: Slip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272138.1 | NP_001259067.1 | 2 - 3 | 604 | 90 | |
| NM_143676.2 | NP_651933.1 | 2 - 3 | 604 | 90 | ||
More Information
Set Membership: |
|

Slip1
Genomic Map