Assay Details
| Gene Symbol: Slip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272137.1 | NP_001259066.1 | 3 - 4 | 1587 | 65 | |
| NM_143676.2 | NP_651933.1 | 4 - 5 | 1905 | 65 | ||
More Information
Set Membership: |
|

Slip1
Genomic Map