Assay Details
| Gene Symbol: DNApol-alpha60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143744.4 | NP_652001.2 | 1 | 1373 | 62 | |
| GenBank mRNA | BT133488.1 | - | 1 | 1317 | 62 | |
More Information
Set Membership: |
|

DNApol-alpha60
Genomic Map