Assay Details
Gene Symbol: deltaCOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169171.2 | NP_001162642.1 | 5 - 6 | 1071 | 64 | |
NM_143755.4 | NP_652012.1 | 5 - 6 | 1074 | 64 | ||
GenBank mRNA | AY051528.1 | - | 5 - 6 | 1076 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
