Assay Details
Gene Symbol: deltaCOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169171.2 | NP_001162642.1 | 7 - 8 | 1492 | 74 | |
NM_143755.4 | NP_652012.1 | 7 - 8 | 1495 | 74 | ||
GenBank mRNA | AY051528.1 | - | 7 - 8 | 1497 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
