Assay Details
| Gene Symbol: ND-SGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274813.1 | NP_001261742.1 | 3 - 4 | 495 | 81 | |
| NM_143785.3 | NP_652042.1 | 3 - 4 | 606 | 81 | ||
| GenBank mRNA | AY070637.1 | - | 3 - 4 | 501 | 81 | |
| AY070682.1 | - | 3 - 4 | 515 | 81 | ||
More Information
Set Membership: |
|

ND-SGDH
Genomic Map