Assay Details
| Gene Symbol: Su(z)12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143802.3 | NP_652059.1 | 5 - 6 | 2892 | 115 | |
| GenBank mRNA | AY069809.1 | - | 5 - 6 | 2830 | 115 | |
More Information
Set Membership: |
|

Su(z)12
Genomic Map