Assay Details
| Gene Symbol: Hipk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274286.1 | NP_001261215.1 | 4 - 5 | 1494 | 98 | |
| NM_001382035.1 | NP_001369036.1 | 5 - 6 | 2173 | 98 | ||
| NM_138194.3 | NP_612038.2 | 5 - 6 | 2095 | 98 | ||
| NM_167834.2 | NP_728531.1 | 5 - 6 | 2173 | 98 | ||
More Information
Set Membership: |
|

Hipk
Genomic Map