Assay Details
| Gene Symbol: Glut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103999.4 | NP_001097469.2 | 3 - 4 | 767 | 92 | |
| NM_001274304.1 | NP_001261233.1 | 8 - 9 | 1401 | 92 | ||
| NM_001274305.1 | NP_001261234.1 | 9 - 10 | 2135 | 92 | ||
| NM_001274306.1 | NP_001261235.1 | 7 - 8 | 1230 | 92 | ||
| NM_001274307.1 | NP_001261236.1 | 6 - 7 | 1209 | 92 | ||
| NM_001274308.1 | NP_001261237.1 | 6 - 7 | 1209 | 92 | ||
| NM_001274309.1 | NP_001261238.1 | 6 - 7 | 1209 | 92 | ||
| NM_167846.3 | NP_728558.5 | 8 - 9 | 1401 | 92 | ||
More Information
Set Membership: |
|

Glut1
Genomic Map