Assay Details
| Gene Symbol: Glut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103999.4 | NP_001097469.2 | 4 - 5 | 910 | 70 | |
| NM_001274305.1 | NP_001261234.1 | 10 - 11 | 2278 | 70 | ||
| NM_001274307.1 | NP_001261236.1 | 7 - 8 | 1352 | 70 | ||
| NM_001274308.1 | NP_001261237.1 | 7 - 8 | 1352 | 70 | ||
| NM_001274309.1 | NP_001261238.1 | 7 - 8 | 1352 | 70 | ||
| NM_167846.3 | NP_728558.5 | 9 - 10 | 1544 | 70 | ||
More Information
Set Membership: |
|

Glut1
Genomic Map