Assay Details
| Gene Symbol: Myo61F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057586.4 | NP_476934.2 | 3 - 4 | 326 | 69 | |
| NM_167870.2 | NP_728594.2 | 3 - 4 | 403 | 69 | ||
| NM_167871.2 | NP_728595.1 | 3 - 4 | 411 | 69 | ||
| NM_167872.2 | NP_728596.1 | 3 - 4 | 549 | 69 | ||
More Information
Set Membership: |
|

Myo61F
Genomic Map