Assay Details
| Gene Symbol: MEP-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144403.2 | NP_001137875.1 | 3 - 4 | 3162 | 93 | |
| NM_139476.3 | NP_647733.1 | 3 - 4 | 2957 | 93 | ||
| NM_167958.2 | NP_728761.1 | 3 - 4 | 2947 | 93 | ||
| NM_167959.2 | NP_728762.1 | 4 - 5 | 3102 | 93 | ||
| NM_167960.2 | NP_728763.1 | 2 - 3 | 2723 | 93 | ||
| NM_167961.2 | NP_728764.1 | 2 - 3 | 2546 | 93 | ||
| NM_167962.2 | NP_728765.1 | 2 - 3 | 2536 | 93 | ||
| Gene Symbol: CG42245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144404.2 | NP_001137876.1 | 4 - 5 | 3102 | 93 | |
| GenBank mRNA | BT023882.1 | - | 4 - 5 | 3097 | 93 | |
More Information
Set Membership: |
|

MEP-1
Genomic Map