Assay Details
| Gene Symbol: MEP-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144403.2 | NP_001137875.1 | 4 - 5 | 3423 | 101 | |
| NM_139476.3 | NP_647733.1 | 4 - 5 | 3218 | 101 | ||
| NM_167958.2 | NP_728761.1 | 4 - 5 | 3208 | 101 | ||
| NM_167959.2 | NP_728762.1 | 5 - 6 | 3363 | 101 | ||
| NM_167960.2 | NP_728763.1 | 3 - 4 | 2984 | 101 | ||
| NM_167961.2 | NP_728764.1 | 3 - 4 | 2807 | 101 | ||
| NM_167962.2 | NP_728765.1 | 3 - 4 | 2797 | 101 | ||
| Gene Symbol: CG42245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144404.2 | NP_001137876.1 | 5 - 6 | 3363 | 101 | |
| GenBank mRNA | BT023882.1 | - | 5 - 6 | 3358 | 101 | |
More Information
Set Membership: |
|

MEP-1
Genomic Map