Assay Details
| Gene Symbol: Drsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168020.3 | NP_728861.1 | 1 | 115 | 77 | |
| GenBank mRNA | AY338361.1 | - | 1 | 77 | 77 | |
| AY351398.1 | - | 1 | 77 | 77 | ||
| KX531450.1 | - | 1 | 77 | 77 | ||
More Information
Set Membership: |
|

Drsl3
Genomic Map