Assay Details
| Gene Symbol: Eip63E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043118.3 | NP_001036583.1 | 2 - 3 | 431 | 110 | |
| NM_001043119.3 | NP_001036584.1 | 3 - 4 | 240 | 110 | ||
| NM_001043120.3 | NP_001036585.1 | 2 - 3 | 459 | 110 | ||
| NM_001043121.2 | NP_001036586.1 | 1 - 2 | 337 | 110 | ||
| NM_001169869.2 | NP_001163340.1 | 1 - 2 | 812 | 110 | ||
| NM_168034.3 | NP_728889.1 | 3 - 4 | 240 | 110 | ||
| NM_168035.2 | NP_728890.1 | 2 - 3 | 459 | 110 | ||
| NM_168036.3 | NP_728891.1 | 2 - 3 | 431 | 110 | ||
| GenBank mRNA | AF152401.1 | - | 2 - 3 | 457 | 110 | |
| AF152402.1 | - | 2 - 3 | 457 | 110 | ||
| AF152404.1 | - | 3 - 4 | 240 | 110 | ||
| AF152405.1 | - | 1 - 2 | 831 | 110 | ||
| AF152406.1 | - | 2 - 3 | 427 | 110 | ||
| BT010064.1 | - | 1 - 2 | 337 | 110 | ||
| BT022123.1 | - | 431 | 110 | |||
More Information
Set Membership: |
|

Eip63E
Genomic Map