Assay Details
| Gene Symbol: TTLL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168100.3 | NP_729025.1 | 1 | 1811 | 67 | |
| GenBank mRNA | BT025844.1 | - | 1 | 1811 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTLL1B| Gene Symbol: TTLL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168100.3 | NP_729025.1 | 1 | 1811 | 67 | |
| GenBank mRNA | BT025844.1 | - | 1 | 1811 | 67 | |
Set Membership: |
|