Assay Details
| Gene Symbol: Fitm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139687.3 | NP_647944.2 | 2 - 3 | 801 | 78 | |
| NM_168110.3 | NP_729053.2 | 1 - 2 | 875 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fitm| Gene Symbol: Fitm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139687.3 | NP_647944.2 | 2 - 3 | 801 | 78 | |
| NM_168110.3 | NP_729053.2 | 1 - 2 | 875 | 78 | ||
Set Membership: |
|