Assay Details
| Gene Symbol: QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168145.3 | NP_729109.1 | 1 - 2 | 123 | 75 | |
| GenBank mRNA | AY119500.1 | - | 1 - 2 | 105 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

QC| Gene Symbol: QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168145.3 | NP_729109.1 | 1 - 2 | 123 | 75 | |
| GenBank mRNA | AY119500.1 | - | 1 - 2 | 105 | 75 | |
Set Membership: |
|