Assay Details
| Gene Symbol: QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168145.3 | NP_729109.1 | 3 - 4 | 523 | 97 | |
| GenBank mRNA | AY119500.1 | - | 3 - 4 | 505 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

QC| Gene Symbol: QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168145.3 | NP_729109.1 | 3 - 4 | 523 | 97 | |
| GenBank mRNA | AY119500.1 | - | 3 - 4 | 505 | 97 | |
Set Membership: |
|