Assay Details
| Gene Symbol: PVRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168153.2 | NP_729124.1 | 4 - 5 | 1432 | 54 | |
| GenBank mRNA | BT010250.1 | - | 4 - 5 | 1425 | 54 | |
More Information
Set Membership: |
|

PVRAP
Genomic Map