Assay Details
| Gene Symbol: PVRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168153.2 | NP_729124.1 | 6 - 7 | 2024 | 91 | |
| GenBank mRNA | BT010250.1 | - | 6 - 7 | 2017 | 91 | |
More Information
Set Membership: |
|

PVRAP
Genomic Map