Assay Details
| Gene Symbol: Dscam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274569.2 | NP_001261498.1 | 21 | 6414 | 82 | |
| NM_001274570.2 | NP_001261499.1 | 21 | 6384 | 82 | ||
| NM_001274571.2 | NP_001261500.1 | 21 | 6414 | 82 | ||
| NM_001316478.1 | NP_001303407.1 | 21 | 6414 | 82 | ||
| NM_168194.4 | NP_729223.2 | 21 | 6396 | 82 | ||
| NM_168196.4 | NP_729225.2 | 21 | 6498 | 82 | ||
More Information
Set Membership: |
|

Dscam2
Genomic Map